Skip to Content
Merck
All Photos(3)

Key Documents

V900708

Sigma-Aldrich

Methyl carbamate

Vetec, reagent grade, 98%

Synonym(s):

Urethylane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COOCH3
CAS Number:
Molecular Weight:
75.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

bp

176-177 °C (lit.)

mp

56-58 °C (lit.)

SMILES string

COC(N)=O

InChI

1S/C2H5NO2/c1-5-2(3)4/h1H3,(H2,3,4)

InChI key

GTCAXTIRRLKXRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P C Chan et al.
Japanese journal of cancer research : Gann, 83(3), 258-263 (1992-03-01)
Short-term and long-term carcinogenicity of methyl carbamate (MCB) was evaluated in F344 rats and B6C3F1 mice. In experiments lasting 6, 12, and 18 months, MCB was given in water by gavage to groups of 10 male and 10 female rats
N P Sen et al.
Food additives and contaminants, 9(2), 149-160 (1992-03-01)
A method is described for the simultaneous determination of methyl carbamate (MC) and ethyl carbamate (EC) in wines that is based on: (a) extraction of the sample with dichloromethane using an extraction tube or an alumina-Celite column, (b) concentration of
Xusheng Shao et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(43), 17273-17277 (2013-10-11)
The nicotinic acetylcholine (ACh) receptor (nAChR) is the principal insecticide target. Nearly half of the insecticides by number and world market value are neonicotinoids acting as nAChR agonists or organophosphorus (OP) and methylcarbamate (MC) acetylcholinesterase (AChE) inhibitors. There was no
John E Casida et al.
Chemico-biological interactions, 203(1), 221-225 (2012-08-29)
The anticholinesterase (antiChE) organophosphorus (OP) and methylcarbamate (MC) insecticides have been used very effectively as contact and systemic plant protectants for seven decades. About 90 of these compounds are still in use - the largest number for any insecticide chemotype
Helen E Carter et al.
Pest management science, 69(9), 1040-1048 (2013-06-14)
Methyl benzimidazole carbamate (MBC) fungicides are used to control the oilseed rape pathogen Pyrenopeziza brassicae. Resistance to MBCs has been reported in P. brassicae, but the molecular mechanism(s) associated with reductions in sensitivity have not been verified in this species.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service