Skip to Content
Merck
All Photos(3)

Key Documents

V900669

Sigma-Aldrich

DL-Valine

Vetec, reagent grade, 99%

Synonym(s):

(±)-α-Aminoisovaleric acid, DL-2-Amino-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NH2)COOH
CAS Number:
Molecular Weight:
117.15
Beilstein:
506689
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

99%

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

colorless to white

mp

295 °C (dec.) (lit.)

SMILES string

CC(C)C(N)C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)

InChI key

KZSNJWFQEVHDMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Hinneh et al.
Food research international (Ottawa, Ont.), 111, 607-620 (2018-07-17)
The practice of pod storage (PS) has been applied in many cocoa producing countries, especially by Ghanaian farmers over the years. However, the study of PS has not received much attention, hence, until now, its potential impact on specific flavor
Yan-Mei Ma et al.
Fish & shellfish immunology, 45(2), 414-421 (2015-05-11)
Streptococcus iniae seriously affects the intensive farming of tilapias. Much work has been conducted on prevention and control of S. iniae infection, but little published information on the metabolic response is available in tilapias against the bacterial infection, and no metabolic
K Aaboe et al.
Diabetes, obesity & metabolism, 17(1), 74-81 (2014-09-23)
To examine whether 12 weeks of treatment with a dipeptidyl peptidase-4 (DPP-4) inhibitor, sitagliptin, influences the insulin secretion induced by glucose, glucose-dependent insulinotropic polypeptide (GIP) and glucagon-like peptide-1 (GLP-1) during a hyperglycaemic clamp in patients with type 2 diabetes (T2DM).
Aleksander Hinek et al.
Journal of dermatological science, 75(3), 173-182 (2014-07-13)
Vitamin C (L-ascorbic acid), a known enhancer of collagen deposition, has also been identified as an inhibitor of elastogenesis. Present studies explored whether and how the L-ascorbic acid derivative (+) sodium L-ascorbate (SA) would affect production of collagen and elastic
Thomas Courivaud et al.
The Journal of biological chemistry, 290(34), 21007-21018 (2015-07-15)
Although E3 ubiquitin ligases are deemed to play key roles in normal cell function and homeostasis, whether their alterations contribute to cancer pathogenesis remains unclear. In this study, we sought to investigate potential mechanisms that govern WWP1/Tiul1 (WWP1) ubiquitin ligase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service