Skip to Content
Merck
All Photos(3)

Documents

V900618

Sigma-Aldrich

4-Hydroxyphenylacetic acid

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
1448766
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

mp

148-151 °C (lit.)

SMILES string

OC(=O)Cc1ccc(O)cc1

InChI

1S/C8H8O3/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4,9H,5H2,(H,10,11)

InChI key

XQXPVVBIMDBYFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth Stensrud et al.
The Journal of organic chemistry, 74(15), 5219-5227 (2009-07-04)
Three new trifluoromethylated p-hydroxyphenacyl (pHP)-caged gamma-aminobutyric acid (GABA) and glutamate (Glu) derivatives have been examined for their efficacy as photoremovable protecting groups in aqueous solution. Through the replacement of hydrogen with fluorine, e.g., a m-trifluoromethyl or a m-trifluoromethoxy versus m-methoxy
S N Young et al.
Journal of neurology, neurosurgery, and psychiatry, 45(7), 633-639 (1982-07-01)
Phenylacetic acid, p-hydroxyphenylacetic acid, m-hydroxyphenylacetic acid, phenylalanine, indoleacetic acid, 5-hydroxyindoleacetic acid and tryptophan were measured in lumbar and cisternal cerebrospinal fluid (CSF) taken during pneumoencephalography. The data suggest that the concentration of the acid metabolites of the trace amines tryptamine
F A Muskiet et al.
Clinical chemistry, 25(7), 1281-1284 (1979-07-01)
We report quantitative data on beta-glucuronidase- and sulfatase-hydrolyzable conjugates of homovanillic acid, 3,4-dihydroxyphenylacetic acid, p-hydroxyphenylacetic acid, and vanillic acid in the urine of 20 apparently normal and healthy control persons and of three patients with neuroblastoma. We used organic solvent
Suri Roowi et al.
Molecular nutrition & food research, 53 Suppl 1, S68-S75 (2009-05-06)
Human urine was collected over a 24 h period after the consumption of 250 mL of (i) water, (ii) orange juice, and (iii) orange juice plus 150 mL of full fat natural yoghurt. The orange juice contained 168 micromol of
Andrew M Jenner et al.
Free radical biology & medicine, 38(6), 763-772 (2005-02-22)
Phenolic compounds are not completely absorbed in the small intestine and so enter the colon, where they might exert physiological effects. To identify phenolics that are present in normal human colon, fecal water was prepared from 5 free-living volunteers with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service