Skip to Content
Merck
All Photos(3)

Key Documents

V900510

Sigma-Aldrich

Agarose

Vetec, reagent grade

Synonym(s):

3,6-Anhydro-α-L-galacto-β-D-galactan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41105317
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei Sun et al.
Frontiers in neural circuits, 14, 53-53 (2020-10-20)
As the most important organ in our bodies, the brain plays a critical role in deciding sex-related differential features; however, the underlying neural circuitry basis remains unclear. Here, we used a cell-type-specific rabies virus-mediated monosynaptic tracing system to generate a
Obayed Raihan et al.
Cell reports, 28(1), 78-90 (2019-07-04)
RNA binding proteins, the key regulators in gene expression at the posttranscriptional level, remain largely uncharacterized with respect to aging and relevant cognitive deterioration. Here, we report that the levels of SFRS11 are substantially decreased in the prefrontal cortex (PFC)
Antoine-Emmanuel Saliba et al.
Nature methods, 11(1), 47-50 (2013-11-26)
Lipids have a role in virtually all biological processes, acting as structural elements, scaffolds and signaling molecules, but they are still largely under-represented in known biological networks. Here we describe a liposome microarray-based assay (LiMA), a method that measures protein
C Caramelo-Nunes et al.
Journal of chromatography. A, 1307, 91-98 (2013-08-07)
Affinity chromatography represents a sole technique in purification of different biomolecules. The specific recognition between affinity ligands and target biomolecules has a major role in the specificity of the process. Therefore, choosing the right ligand is a crucial step for
Michael J Lafferty et al.
The Journal of biological chemistry, 288(40), 28524-28534 (2013-08-21)
Elevated triglycerides are associated with an increased risk of cardiovascular disease, and lipoprotein lipase (LPL) is the rate-limiting enzyme for the hydrolysis of triglycerides from circulating lipoproteins. The N-terminal domain of angiopoietin-like protein 4 (ANGPTL4) inhibits LPL activity. ANGPTL4 was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service