Skip to Content
Merck
All Photos(3)

Key Documents

V900483

Sigma-Aldrich

Trizma® base

≥99%, crystalline powder, reagent grade, Vetec

Synonym(s):

2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
Pricing and availability is not currently available.

Product Name

Trizma® base, Vetec, reagent grade, ≥99%

grade

reagent grade

description

aminopeptidase substrate

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

10.5-12

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)

solubility

water: 0.667 g/mL, clear, colorless to faintly yellow

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trizma® base is a basimetric standard used in biochemistry and molecular biology. It may be used as a buffer or as a component of mixed buffer formulations. It is essentially stable, relatively non-hygroscopic and has a high equivalent weight.[1][2]

Application

Trizma® base can be used as a buffer:
  • In the evanescent absorption analysis of NADPH.[3]
  • In the autoxidation studies of oxymyoglobin.[4]

Other Notes

The pH values of all buffers are temperature- and concentration-dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany
Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Autoxidation of native oxymyoglobin. Kinetic analysis of the pH profile
Shikama K and Sugawara Y
European Journal of Biochemistry, 91(2), 407-413 (1978)
Preparation of buffers for use in enzyme studies
Handbook of Biochemistry and Molecular Biology (2010)
Tapered fibers for cell studies
Reviews in Fluorescence (2005)
Tri (hydroxymethyl) aminomethane as Acidimetric Standard.
Fossum J, et al.
Analytical Chemistry, 23(3), 491-493 (1951)
Corey A Baron et al.
Magnetic resonance in medicine, 73(3), 1075-1084 (2014-04-12)
An acquisition method that does not increase scan time or specific absorption rate is investigated for reducing the deleterious effects of cerebrospinal fluid (CSF) partial volume effects on diffusion tensor imaging (DTI) tractography. It is based on using a shorter

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service