Skip to Content
Merck
All Photos(3)

Documents

V900470

Sigma-Aldrich

L-Tryptophan

≥98%, Vetec

Synonym(s):

(S)-2-Amino-3-(3-indolyl)propionic acid, L-α-Amino-3-indolepropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
Beilstein:
86197
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

product name

L-Tryptophan, Vetec, reagent grade, ≥98%

grade

reagent grade

product line

Vetec

Assay

≥98%

form

powder

color

white to off-white

mp

280-285 °C (dec.) (lit.)

SMILES string

N[C@@H](Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1

InChI key

QIVBCDIJIAJPQS-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Amino acid precursor of serotonin and melatonin

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kei Hiruma et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(23), 9589-9594 (2013-05-23)
The hypersensitive response (HR) is a type of strong immune response found in plants that is accompanied by localized cell death. However, it is unclear how HR can block a broad range of pathogens with different infective modes. In this
Dirk E M Geurts et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(48), 18932-18939 (2013-11-29)
Adaptive decision-making involves interaction between systems regulating Pavlovian and instrumental control of behavior. Here we investigate in humans the role of serotonin in such Pavlovian-instrumental transfer in both the aversive and the appetitive domain using acute tryptophan depletion, known to
Michael Platten et al.
Cancer research, 72(21), 5435-5440 (2012-10-24)
Tryptophan catabolism in cancer is increasingly being recognized as an important microenvironmental factor that suppresses antitumor immune responses. It has been proposed that the essential amino acid tryptophan is catabolized in the tumor tissue by the rate-limiting enzyme indoleamine-2,3-dioxygenase (IDO)
Annemieke T van der Goot et al.
Trends in molecular medicine, 19(6), 336-344 (2013-04-09)
Aging is an important risk factor for many debilitating diseases, including cancer and neurodegeneration. In model organisms, interfering with metabolic signaling pathways, including the insulin/insulin-like growth factor (IGF) 1 (IIS) and TOR pathways, can protect against age-related pathologies and increase
Alexander A Voityuk et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(14), 5219-5224 (2014-03-19)
UV-B absorption by the photoreceptor UV resistance locus 8 (UVR8) consisting of two identical protein units triggers a signal chain used by plants in connection with protection and repair of UV-B induced damage. X-ray structural analysis of the purified protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service