Skip to Content
Merck
All Photos(2)

Key Documents

V900458

Sigma-Aldrich

L-Asparagine

98%, Vetec

Synonym(s):

(S)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
132.12
Beilstein:
1723527
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

product name

L-Asparagine, Vetec, reagent grade, 98%

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

color

white

mp

235 °C (dec.) (lit.)

SMILES string

N[C@@H](CC(N)=O)C(O)=O

InChI

1S/C4H8N2O3/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H2,6,7)(H,8,9)/t2-/m0/s1

InChI key

DCXYFEDJOCDNAF-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuo Yanagisawa et al.
Acta crystallographica. Section D, Biological crystallography, 69(Pt 1), 5-15 (2013-01-01)
Structures of Methanosarcina mazei pyrrolysyl-tRNA synthetase (PylRS) have been determined in a novel crystal form. The triclinic form crystals contained two PylRS dimers (four monomer molecules) in the asymmetric unit, in which the two subunits in one dimer each bind
Christopher J Poon et al.
The Journal of biological chemistry, 280(25), 23615-23621 (2005-04-26)
Keratan sulfate is thought to influence the cleavage of aggrecan by metalloenzymes. We have therefore produced a recombinant substrate, substituted with keratan sulfate, suitable for the study of aggrecanolysis in vitro. Recombinant human G1-G2 was produced in primary bovine keratocytes
Alberto Canepa et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 17(3), 413-421 (2002-02-28)
The aims of this study were to investigate free amino acid (AA) concentrations in plasma, red blood cells (RBC), polymorphonuclear granulocytes (PMN), and muscle, sampled at the same time, in normal and uraemic children. Twelve apparently well-nourished chronically uraemic children
Emma R Job et al.
Journal of immunology (Baltimore, Md. : 1950), 190(5), 2169-2177 (2013-02-01)
Seasonal influenza A viruses (IAV) originate from pandemic IAV and have undergone changes in antigenic structure, including addition of glycans to the viral hemagglutinin (HA). Glycans on the head of HA promote virus survival by shielding antigenic sites, but highly
B Ahlman et al.
Clinical science (London, England : 1979), 86(6), 653-662 (1994-06-01)
1. The effects of short-term starvation and refeeding on the free amino acid concentrations of the intestinal mucosa were characterized in male subjects (n = 6), using endoscopically obtained biopsy specimens from the duodenum and from all four segments of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service