Skip to Content
Merck
All Photos(2)

Key Documents

V900437

Sigma-Aldrich

Thymine

Vetec, reagent grade, 99%

Synonym(s):

2,4-Dihydroxy-5-methylpyrimidine, 5-Methyluracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
Beilstein:
117880
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

99%

mp

~320 °C (dec.) (lit.)

SMILES string

CC1=CNC(=O)NC1=O

InChI

1S/C5H6N2O2/c1-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)

InChI key

RWQNBRDOKXIBIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefano Stella et al.
Acta crystallographica. Section D, Biological crystallography, 69(Pt 9), 1707-1716 (2013-09-04)
Transcription activator-like effectors contain a DNA-binding domain organized in tandem repeats. The repeats include two adjacent residues known as the repeat variable di-residue, which recognize a single base pair, establishing a direct code between the dipeptides and the target DNA.
Subhendu Sekhar Bag et al.
The Journal of organic chemistry, 78(2), 278-291 (2012-11-23)
We report the design and synthesis of triazolyl donor/acceptor unnatural nucleosides via click chemistry and studies on the duplex stabilization of DNA containing two such new nucleosides. The observed duplex stabilization among the self-pair/heteropair has been found to be comparable
U Hardeland et al.
Progress in nucleic acid research and molecular biology, 68, 235-253 (2001-09-14)
More than 50% of colon cancer-associated mutations in the p53 tumor suppressor gene are C-->T transitions. The majority of them locate in CpG dinucleotides and are thought to have arisen through spontaneous hydrolytic deamination of 5-methylcytosine. This deamination process gives
S I Ahmad et al.
Annual review of microbiology, 52, 591-625 (1999-01-19)
For many years it has been known that thymine auxotrophic microorganisms undergo cell death in response to thymine starvation [thymineless death (TLD)]. This effect is unusual in that deprivation of many other nutritional requirements has a biostatic, but not lethal
M Weinfeld et al.
Progress in nucleic acid research and molecular biology, 68, 139-149 (2001-09-14)
The toxic and mutagenic effects of ionizing radiation are believed to be caused by damage to cellular DNA. We have made use of a novel immunoassay for thymine glycol to examine the removal of this lesion from the DNA of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service