V900355
3,3′,5,5′-Tetramethylbenzidine dihydrochloride hydrate
Vetec™, reagent grade, ≥98%
Synonym(s):
TMB dihydrochloride
About This Item
grade
reagent grade
product line
Vetec™
Assay
≥98%
solubility
acetic acid: water (1:2): 0.1 g/5mL, clear to slightly hazy
SMILES string
O.Cl.Cl.Cc1cc(cc(C)c1N)-c2cc(C)c(N)c(C)c2
InChI
1S/C16H20N2.2ClH.H2O/c1-9-5-13(6-10(2)15(9)17)14-7-11(3)16(18)12(4)8-14;;;/h5-8H,17-18H2,1-4H3;2*1H;1H2
InChI key
KVCWTKDFVVSVSJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 4 - Carc. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service