Skip to Content
Merck
All Photos(3)

Key Documents

V900343

Sigma-Aldrich

L-Arginine

≥98%, Vetec

Synonym(s):

(S)-2-Amino-5-guanidinopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NH(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
174.20
Beilstein:
1725413
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

Product Name

L-Arginine, Vetec, reagent grade, ≥98%

grade

reagent grade

product line

Vetec

Assay

≥98%

form

powder

color

white

mp

222 °C (dec.) (lit.)

SMILES string

N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1

InChI key

ODKSFYDXXFIFQN-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • L-Arginine-Dependent Nitric Oxide Production in the Blood of Patients with Type 2 Diabetes: A Pilot, Five-Year Prospective Study.: This study examines the role of L-Arginine in nitric oxide production in diabetic patients, proposing its potential benefits for managing vascular complications in diabetes (Stoian et al., 2024).

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Diane N Singhroy et al.
Retrovirology, 10, 73-73 (2013-07-23)
Protein arginine methyltransferase 6 (PRMT6) is a nuclear enzyme that methylates arginine residues on histones and transcription factors. In addition, PRMT6 inhibits HIV-1 replication in cell culture by directly methylating and interfering with the functions of several HIV-1 proteins, i.e.
Ahmed Moursy et al.
Nucleic acids research, 42(10), 6659-6672 (2014-04-03)
Regulation of SMN2 exon 7 splicing is crucial for the production of active SMN protein and the survival of Spinal Muscular Atrophy (SMA) patients. One of the most efficient activators of exon 7 inclusion is hnRNP G, which is recruited
Kaoru Kumazaki et al.
Nature, 509(7501), 516-520 (2014-04-18)
Newly synthesized membrane proteins must be accurately inserted into the membrane, folded and assembled for proper functioning. The protein YidC inserts its substrates into the membrane, thereby facilitating membrane protein assembly in bacteria; the homologous proteins Oxa1 and Alb3 have
Hartmut Grasemann et al.
Current pharmaceutical design, 18(5), 726-736 (2012-01-11)
The concentrations of nitric oxide are decreased in airways of patients with cystic fibrosis. The reasons for this nitric oxide deficiency are incompletely understood but may include reduced production from nitric oxide synthases due to decreased expression, the enzymes in
John Pernow et al.
Cardiovascular research, 98(3), 334-343 (2013-02-19)
Functional integrity of the vascular endothelium is of fundamental importance for normal vascular function. A key factor regulating endothelial function is the bioavailability of nitric oxide (NO). Recently, the enzyme arginase has emerged as an important regulator of NO production

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service