Skip to Content
Merck
All Photos(2)

Documents

V900316

Sigma-Aldrich

6-Benzylaminopurine

Vetec, reagent grade, 98%

Synonym(s):

6-BAP, BA, N6-Benzyladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N5
CAS Number:
Molecular Weight:
225.25
Beilstein:
19406
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

application(s)

agriculture

SMILES string

C(Nc1ncnc2nc[nH]c12)c3ccccc3

InChI

1S/C12H11N5/c1-2-4-9(5-3-1)6-13-11-10-12(15-7-14-10)17-8-16-11/h1-5,7-8H,6H2,(H2,13,14,15,16,17)

InChI key

NWBJYWHLCVSVIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Feng Xu et al.
Journal of agricultural and food chemistry, 60(1), 234-240 (2011-12-14)
The effect of 6-benzylaminopurine (6-BA) on the color, antioxidant activity, and contents of total phenols, glucosinolate, and sulforaphane in broccoli florets was investigated. The results showed that 6-BA treatment markedly inhibited the increase of the L* value and malondialdehyde (MDA)
Bijesh Puthusseri et al.
Plant foods for human nutrition (Dordrecht, Netherlands), 67(2), 162-170 (2012-04-12)
Folate (vitamin B₉) content was evaluated in 10 varieties of coriander with the aim of enhancing its concentration and stability, because of three reasons: 1) coriander is among a few widely used greens in the world and suits many cuisines
Zdeněk Trávníček et al.
Journal of inorganic biochemistry, 105(7), 937-948 (2011-05-04)
A series of novel octahedral ruthenium(III) complexes involving 6-benzylaminopurine (L) derivatives as N-donor ligands has been prepared by the reaction of [(DMSO)(2)H][trans-RuCl(4)(DMSO)(2)] with the corresponding L derivative. The complexes 1-12 have the general compositions trans-[RuCl(4)(DMSO)(n-Cl-LH)]⋅xSol (1-3), trans-[RuCl(4)(DMSO)(n-Br-LH)]·xSol (4-6), trans-[RuCl(4)(DMSO)(n-OMe-LH)]·xSol (7-9)
Anne Birgitte Heckmann et al.
Molecular plant-microbe interactions : MPMI, 24(11), 1385-1395 (2011-07-21)
Cytokinin plays a central role in the formation of nitrogen-fixing root nodules following inoculation with rhizobia. We show that exogenous cytokinin induces formation of discrete and easily visible nodule primordia in Lotus japonicus roots. The expression of nodulin genes was
Alexandar Tzankov et al.
Journal of hematology & oncology, 8, 70-70 (2015-06-14)
The prognostic role of tumor-related parameters in diffuse large B cell lymphoma (DLBCL) is a matter of controversy. We investigated the prognostic value of phenotypic and genotypic profiles in DLBCL in clinical trial (NCT00544219) patients homogenously treated with six cycles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service