Skip to Content
Merck
All Photos(1)

Key Documents

V900281

Sigma-Aldrich

4-(Dimethylamino)benzaldehyde

Vetec, reagent grade, 98%

Synonym(s):

Ehrlich’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4CHO
CAS Number:
Molecular Weight:
149.19
Beilstein:
606802
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

mp

72-75 °C (lit.)

SMILES string

CN(C)c1ccc(C=O)cc1

InChI

1S/C9H11NO/c1-10(2)9-5-3-8(7-11)4-6-9/h3-7H,1-2H3

InChI key

BGNGWHSBYQYVRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Forms colored condensation products (Schiff bases) with pyrroles and primary amines.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

327.2 °F - closed cup

Flash Point(C)

164 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Creuzenet et al.
The Journal of biological chemistry, 275(45), 34873-34880 (2000-08-24)
FlaA1 is a small soluble protein of unknown function in Helicobacter pylori. It has homologues that are essential for the virulence of numerous medically relevant bacteria. FlaA1 was overexpressed as a histidine-tagged protein and purified to homogeneity by nickel chelation
M Stefek et al.
Biochimica et biophysica acta, 1502(3), 398-404 (2000-11-09)
In the present work, pepsin digests of tail tendons from streptozotocin-diabetic rats were found to contain material that reacted rapidly at room temperature with p-dimethylaminobenzaldehyde (Ehrlich's reagent) to give an adduct with an absorbance spectrum characteristic of the Ehrlich chromogen
Rosario Zamora et al.
Journal of agricultural and food chemistry, 52(13), 4166-4171 (2004-06-24)
The Ehrlich reaction was optimized to determine the formation of pyrrolized phospholipids in edible oils in an attempt to understand the color reversion produced during the deodorization of poorly degummed edible oils. The procedure consisted of the treatment of the
G P Kushto et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 54A(6), 799-819 (1998-07-17)
Results of a normal coordinate analysis based on infrared and Raman spectra of six isotopic forms of 4-(dimethylamino)benzaldehyde are reported (DABA, DABA-CDO, DABA-13CDO, DABA-13CHO, DABA-CHO18O, and DABA-3,5-D). Cs point group symmetry has been applied and all motions except the internal
Jun-Min Guo et al.
Journal of agricultural and food chemistry, 58(11), 6556-6561 (2010-05-15)
A simple colorimetric method for the differentiation of indoleacetic acid (IAA) and indolebutyric acid (IBA) in plant samples is described. The color change is based upon the reaction between the auxins and p-(dimethylamino)benzaldehyde (PDAB, Ehrlich reagent) following the electrophilic substitution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service