Skip to Content
Merck
All Photos(3)

Key Documents

V900265

Sigma-Aldrich

Sodium phosphate monobasic monohydrate

Vetec, reagent grade, 98%

Synonym(s):

Monosodium phosphate, Sodium dihydrogen phosphate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4 · H2O
CAS Number:
Molecular Weight:
137.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
98%
grade:
reagent grade
form:
crystalline
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

98%

form

crystalline

pH

4.1-4.5 (25 °C, 50 g/L)

SMILES string

[Na+].[H]O[H].OP(O)([O-])=O

InChI

1S/Na.H3O4P.H2O/c;1-5(2,3)4;/h;(H3,1,2,3,4);1H2/q+1;;/p-1

InChI key

BBMHARZCALWXSL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehrnoosh Shirangi et al.
Pharmaceutical research, 32(9), 3044-3054 (2015-04-03)
Polyesters with hydrophilic domains, i.e., poly(D,L-lactic-co-glycolic-co-hydroxymethyl glycolic acid) (PLGHMGA) and a multiblock copolymer of poly(ε-caprolactone)-PEG-poly(ε-caprolactone) and poly(L-lactide) ((PC-PEG-PC)-(PL)) are expected to cause less acylation of encapsulated peptides than fully hydrophobic matrices. Our purpose is to assess the extent and sites
Christopher Polonyi et al.
Dalton transactions (Cambridge, England : 2003), 43(31), 11941-11949 (2014-06-27)
The ability of the anticancer active drug cisplatin to exert biological activity through interference with nucleic acid function is well documented. Since kinetics play a key role in determining product distributions in these systems, methods for accurate documentation of reactivity
Genye He et al.
Journal of chromatographic science, 52(8), 848-851 (2013-09-14)
Formoterol is a new threshold substance in the prohibited list 2012 according to World Anti-Doping Agency. Extracted by ethyl acetate using formoterol-D6 as internal standard, formoterol underwent a constant flow rate gradient elution separation in reversed-phase liquid chromatography. Subsequently, mass
C Fornaguera et al.
Nanoscale, 7(28), 12076-12084 (2015-06-30)
Polymeric nanoparticles could be promising drug delivery systems to treat neurodegenerative diseases. Among the various methods of nanoparticle preparation, nano-emulsion templating was used in the present study to prepare galantamine-loaded nano-emulsions by a low-energy emulsification method followed by solvent evaporation
Sry D Hujaya et al.
Acta biomaterialia, 22, 19-31 (2015-05-02)
Dip-coated multilayered thin films of poly(amido amine)s (PAAs) and DNA have been developed to provide surfaces with cell-transfecting capabilities. Three types of PAAs, differing in side chain functional groups, were synthesized and characterized for their properties in forming multilayered structures

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service