Skip to Content
Merck
All Photos(2)

Key Documents

V900157

Sigma-Aldrich

Ethylenediaminetetraacetic acid dipotassium salt dihydrate

Vetec, reagent grade, 98%

Synonym(s):

(Ethylenedinitrilo)tetraacetic acid dipotassium salt, Dipotassium ethylenediaminetetraacetate dihydrate, EDTA dipotassium salt, Edathamil, Potassium ethylenediaminetetraacetate dibasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2(KOOCCH2)NCH2CH2N(CH2COOK)CH2COOH · 2H2O
CAS Number:
Molecular Weight:
404.45
EC Number:
UNSPSC Code:
12352107
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

mp

255-280 °C (dec.) (lit.)

solubility

3 M KOH: 160 mg/mL, clear to very slightly hazy, colorless to faintly yellow

SMILES string

O.O.O.O.[K+].[K+].[K+].[K+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2K.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

JFROQFOCFOKDKU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa K Philp et al.
PloS one, 10(2), e0117494-e0117494 (2015-02-07)
High saturated fat (HF-S) diets increase intramyocellular lipid, an effect ameliorated by omega-3 fatty acids in vitro and in vivo, though little is known about sex- and muscle fiber type-specific effects. We compared effects of standard chow, HF-S, and 7.5%
Anne-Marie L Seymour et al.
Cardiovascular research, 106(2), 249-260 (2015-03-10)
Left ventricular hypertrophy is an adaptive response of the heart to chronic mechanical overload and can lead to functional deterioration and heart failure. Changes in cardiac energy metabolism are considered as key to the hypertrophic remodelling process. The concurrence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service