Skip to Content
Merck
All Photos(3)

Key Documents

V900156

Sigma-Aldrich

Poly(ethylene glycol)

Vetec, reagent grade, avg mol wt 8,000

Synonym(s):

PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352100

grade

reagent grade

product line

Vetec

mol wt

avg mol wt 8,000

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyethylene glycol (PEG) is a synthetic, hydrophilic, biocompatible polymer that has a wide range of uses in PEGylation, surface conjugation, nanoparticle coating, and crosslinking for hydrogels, etc. PEGs are made by polymerizing ethylene oxide via a ring-opening reaction.

Application

Poly(ethylene glycol) avg mol wt 8,000 can be used as:      
  • A stabilizing agent in the synthesis of silver nanoparticles via solid-state oxidation of silver nitrate.     
  • A reaction medium in the synthesis of dibenz[b,f]-1,4-oxazepine via base-catalyzed cyclization of 2-fluoro benzaldehyde and 2-amino phenol.      
  • A surfactant to prepare PEGylated CuO nanoparticles via hydrothermal method using Cu(NO3)2 as a precursor salt.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Giannousi et al.
Journal of inorganic biochemistry, 164, 82-90 (2016-09-26)
There is a growing field of research into the physicochemical properties of metal oxide nanoparticles (NPs) and their potential use against tumor formation, development and progression. Coated NPs with biocompatible surfactants can be incorporated into the natural metabolic pathway of
D Law et al.
Journal of pharmaceutical sciences, 90(8), 1015-1025 (2001-09-06)
A systematic study of the properties of ritonavir and the influence of polyethylene glycol 8000 (PEG) on ritonavir revealed that amorphous ritonavir dispersions in PEG would have an improved dissolution profile and could exhibit long-term stability. Ritonavir, a human immunodeficiency
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced
D D Smyth et al.
Cardiovascular drugs and therapy, 4(1), 297-300 (1990-02-01)
Previous studies have demonstrated that Separan AP-30, a drag-reducing polymer, significantly decreased the formation of atherosclerotic plaques in rabbits fed a high-cholesterol diet. Furthermore, Separan AP-273, a polymer similar to but longer than Separan AP-30, markedly increased cardiac output in
M Patel Geeta et al.
Current drug delivery, 6(2), 159-165 (2009-05-20)
Carbamazepine indicated for the control of epilepsy, undergoes extensive hepatic first-pass metabolism after oral administration. A vaginal dosage form of carbamazepine is not commercially available. Conventional suppository having poor retention in the vaginal tract, as they are removed in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service