Skip to Content
Merck
All Photos(3)

Key Documents

V900086

Sigma-Aldrich

Thioacetamide

Vetec, reagent grade, 98%

Synonym(s):

Ethanethioamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CSNH2
CAS Number:
Molecular Weight:
75.13
Beilstein:
506006
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

mp

108-112 °C (lit.)

functional group

amine

SMILES string

CC(N)=S

InChI

1S/C2H5NS/c1-2(3)4/h1H3,(H2,3,4)

InChI key

YUKQRDCYNOVPGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Zhan et al.
Archives of pharmacal research, 35(6), 975-986 (2012-08-09)
In continuation of our endeavor to develop new, potent, selective and less toxic antiviral agents, a novel series of 2-(2-amino/chloro-4-(2,4-dibromophenyl) thiazol-5-ylthio)acetamide derivatives was synthesized via an expeditious route and evaluated for their anti-HIV activities against wild-type virus and clinically relevant
Chiung-Kuei Huang et al.
Hepatology (Baltimore, Md.), 57(4), 1550-1563 (2012-11-15)
Transplantation of bone marrow mesenchymal stem cells (BM-MSCs) has been considered as an alternative therapy, replacing liver transplantation in clinical trials, to treat liver cirrhosis, an irreversible disease that may eventually lead to liver cancer development. However, low survival rate
M Jamshidi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 153, 257-267 (2015-09-01)
In this work, central composite design (CCD) combined with response surface methodology (RSM) and desirability function approach (DFA) gives useful information about operational condition and also to obtain useful information about interaction and main effect of variables concerned to simultaneous
Hong Ling et al.
PloS one, 8(1), e54499-e54499 (2013-01-26)
Considerable evidence has demonstrated that transforming growth factor β (TGF-β) plays a key role in hepatic fibrosis, the final common pathway for a variety of chronic liver diseases leading to liver insufficiency. Although a few studies have reported that blocking
Heather Hajovsky et al.
Chemical research in toxicology, 25(9), 1955-1963 (2012-08-08)
The hepatotoxicity of thioacetamide (TA) has been known since 1948. In rats, single doses cause centrolobular necrosis accompanied by increases in plasma transaminases and bilirubin. To elicit these effects, TA requires oxidative bioactivation, leading first to its S-oxide (TASO) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service