Skip to Content
Merck
All Photos(3)

Key Documents

V900020

Sigma-Aldrich

Magnesium chloride hexahydrate

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
reagent grade
form:
solid

grade

reagent grade

product line

Vetec

Assay

98%

form

solid

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Compounding MgCl₂·6H₂O with NH₄Al(SO₄)₂·12H₂O or KAl(SO₄)₂·12H₂O to Obtain Binary Hydrated Salts as High-Performance Phase Change Materials.: This research investigates the synthesis and performance of binary hydrated salts combining magnesium chloride hexahydrate with other compounds, highlighting their potential as phase change materials (Sun W et al., 2019).


  • Main chemical species and molecular structure of deep eutectic solvent studied by experiments with DFT calculation: a case of choline chloride and magnesium chloride hexahydrate.: The study delves into the molecular structure and chemical species of a deep eutectic solvent composed of choline chloride and magnesium chloride hexahydrate, using experimental data and DFT calculations (Zhang C et al., 2014).


  • Preparation of extracts for electrophoresis from citrus leaves.: This article presents a method for preparing extracts from citrus leaves for electrophoresis, with magnesium chloride hexahydrate playing a key role in the extraction process (King BJ et al., 1994).


  • Threshold to N-methyl-D-aspartate-induced seizures in mice undergoing chronic nutritional magnesium deprivation is lowered in a way partly responsive to acute magnesium and antioxidant administrations.: This research examines the effects of chronic magnesium deprivation and subsequent magnesium chloride supplementation on seizure thresholds in mice, providing insights into magnesium′s neuroprotective properties (Maurois P et al., 2009).


Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xingchu Gong et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 93, 260-266 (2015-04-25)
Brittleness is one of the important material properties that influences the success or failure of powder compaction. We have discovered that the reciprocal of diametrical elastic strain at fracture is the most suitable tablet brittleness indices (TBIs) for quantifying brittleness
Liang Yang et al.
International journal of pharmaceutics, 471(1-2), 258-263 (2014-05-20)
Telmisartan (TEL) belongs to BCS class II (low solubility/high permeability) and exhibits the pH-dependent drug release. Since 3-aminopropyl functionalized magnesium phyllosilicate (aminoclay) can intercalate or adsorb the negatively charged molecules via the electrostatic interaction, TEL-aminoclay complex was synthesized to improve
Monica Mazzarino et al.
Analytica chimica acta, 837, 70-82 (2014-07-09)
This article describes a liquid chromatographic/tandem mass spectrometric method, based on the use of precursor ion scan as the acquisition mode, specifically developed to detect indole-derived cannabinoids (phenylacetylindoles, naphthoylindoles and benzoylindoles) in biological fluids (saliva, urine and blood). The method
Ikuo Yamamiya et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(9), 1485-1492 (2014-07-09)
Tegafur (FT), a prodrug of 5-fluorouracil, is a chiral molecule, a racemate of R- and S-isomers, and CYP2A6 plays an important role in the enantioselective metabolism of FT in human liver microsomes (R-FT > S-FT). This study examined the enantioselective
Heleen Kraan et al.
Vaccine, 33(17), 2030-2037 (2015-03-17)
In the near future oral polio vaccine (OPV) will be replaced by inactivated polio vaccine (IPV) as part of the eradication program of polio. For that reason, there is a need for substantial amount of safe and more affordable IPV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service