Skip to Content
Merck
All Photos(1)

Key Documents

V800441

Sigma-Aldrich

Urea

AR, ≥99%

Synonym(s):

Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

AR

product line

Vetec

Assay

≥99%

mp

132-135 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gurudayal et al.
Nano letters, 15(6), 3833-3839 (2015-05-06)
Photoelectrochemical water splitting half reactions on semiconducting photoelectrodes have received much attention but efficient overall water splitting driven by a single photoelectrode has remained elusive due to stringent electronic and thermodynamic property requirements. Utilizing a tandem configuration wherein the total
Florin Sasarman et al.
Human molecular genetics, 24(10), 2841-2847 (2015-02-06)
Addition of the trinucleotide cytosine/cytosine/adenine (CCA) to the 3' end of transfer RNAs (tRNAs) is essential for translation and is catalyzed by the enzyme TRNT1 (tRNA nucleotidyl transferase), which functions in both the cytoplasm and mitochondria. Exome sequencing revealed TRNT1
L Li et al.
Cell death and differentiation, 22(7), 1081-1093 (2014-12-20)
P53 is critically important in preventing oncogenesis but its role in inflammation in general and in the function of inflammatory macrophages in particular is not clear. Here, we show that bone marrow-derived macrophages exhibit endogenous p53 activity, which is increased
G L French et al.
Lancet (London, England), 2(8551), 117-119 (1987-07-18)
Tuberculostearic acid, a structural component of Mycobacterium tuberculosis, was identified by gas chromatography/mass spectrometry with selected ion monitoring in cerebrospinal fluid (CSF) from 13 patients with proven and 8 out of 9 patients with suspected tuberculous meningitis; the negative result
Annette Bahlinger et al.
Angewandte Chemie (International ed. in English), 53(33), 8779-8783 (2014-03-20)
β-Amino thioesters are important natural building blocks for the synthesis of numerous bioactive molecules. An organocatalyzed Mannich reaction was developed which provides direct and highly stereoselective access to acyclic β(2)- and β(2,3,3)-amino thioesters with adjacent tertiary and quaternary stereocenters. Mechanistic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service