Skip to Content
Merck
All Photos(2)

Key Documents

V800376

Sigma-Aldrich

Sodium phosphate monobasic

AR, ≥98%

Synonym(s):

Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4
CAS Number:
Molecular Weight:
119.98
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥98%
grade:
AR
form:
powder

grade

AR

product line

Vetec

Assay

≥98%

form

powder

pH

4.0- 4.5 (25 °C, 50 g/L)

cation traces

Fe: ≤0.001%
K: ≤0.01%
heavy metals (as Pb): ≤0.001%

SMILES string

[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

AJPJDKMHJJGVTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Adler et al.
Gastrointestinal endoscopy, 76(6), 1170-1174 (2012-10-03)
Colon capsule endoscopy (CCE) is a noninvasive technique for the detection of colorectal lesions. However, for CCE to be offered as an out-of-clinic procedure, the system needs to automatically alert the patient when to ingest the laxative (booster). We tested
Charles W Kimbrough et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(20), 4576-4585 (2015-07-01)
pH-low insertion peptides (pHLIP) can serve as a targeting moiety that enables pH-sensitive probes to detect solid tumors. Using these probes in conjunction with multispectral optoacoustic tomography (MSOT) is a promising approach to improve imaging for pancreatic cancer. A pH-sensitive
Johanna Troberg et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(1), 107-118 (2014-10-11)
Understanding drug glucuronidation in the dog, a preclinical animal, is important but currently poorly characterized at the level of individual enzymes. We have constructed cDNAs for the 10 dog UDP-glucuronosyltransferases of subfamily 1A (dUGT1As), expressed them in insect cells, and
Samira Hosseini et al.
Biosensors & bioelectronics, 69, 257-264 (2015-03-15)
Electrospun polyhydroxybutyrate (PHB) fibers were dip-coated by polymethyl methacrylate-co-methacrylic acid, poly(MMA-co-MAA), which was synthesized in different molar ratios of the monomers via free-radical polymerization. Fabricated platfrom was employed for immobilization of the dengue antibody and subsequent detection of dengue enveloped
Sima Rahimian et al.
Biomaterials, 61, 33-40 (2015-05-21)
This study investigated the feasibility of the use of polymeric microparticles for sustained and local delivery of immunomodulatory antibodies in immunotherapy of cancer. Local delivery of potent immunomodulatory antibodies avoids unwanted systemic side effects while retaining their anti-tumor effects. Microparticles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service