Skip to Content
Merck
All Photos(2)

Documents

V800359

Sigma-Aldrich

Silver nitrate

LR, ≥99.5%

Synonym(s):

Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

LR

vapor density

5.8 (vs air)

product line

Vetec

Assay

≥99.5%

form

solid

mp

212 °C (dec.) (lit.)

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Hebeish et al.
Carbohydrate polymers, 92(1), 407-413 (2012-12-12)
Innovative CMC-based hydrogels with great potentials for usage in medical area were principally synthesized as per two strategies .The first involved reaction of epichlorohydrin in alkaline medium containing silver nitrate to yield silver nano-particles (AgNPs)-loaded CMC hydrogel. While CMC acted
Liming Wang et al.
ACS nano, 9(6), 6532-6547 (2015-05-23)
To predict potential medical value or toxicity of nanoparticles (NPs), it is necessary to understand the chemical transformation during intracellular processes of NPs. However, it is a grand challenge to capture a high-resolution image of metallic NPs in a single
Tao Xu et al.
Organic letters, 14(21), 5416-5419 (2012-10-24)
A silver-catalyzed intramolecular oxidative aminofluorination of alkynes has been developed by using NFSI as a fluorinating reagent. This reaction represents an efficient method for the synthesis of various 4-fluoroisoquinolines and 4-fluoropyrrolo[α]isoquinolines.
Benjamin P Colman et al.
PloS one, 8(2), e57189-e57189 (2013-03-08)
A large fraction of engineered nanomaterials in consumer and commercial products will reach natural ecosystems. To date, research on the biological impacts of environmental nanomaterial exposures has largely focused on high-concentration exposures in mechanistic lab studies with single strains of
Irina Blinova et al.
Environmental science and pollution research international, 20(5), 3456-3463 (2012-11-13)
Although silver nanoparticles (NPs) are increasingly used in various consumer products and produced in industrial scale, information on harmful effects of nanosilver to environmentally relevant organisms is still scarce. This paper studies the adverse effects of silver NPs to two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service