V800319
Potassium ferricyanide(III)
LR, ≥97%
Synonym(s):
Potassium hexacyanoferrate(III), Red prussiate
About This Item
Recommended Products
grade
LR
product line
Vetec™
Assay
≥97%
form
crystalline
pH
6.0-9.0 (25 °C, 329 g/L)
anion traces
chloride (Cl-): ≤0.05%
hexacyanoferrate(III) ([Fe(CN)6]3-): <0.15%
SMILES string
[K+].[K+].[K+].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N
InChI
1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1
InChI key
MIMJFNVDBPUTPB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- In the preparation of RAFT agent (used in RAFT polymerization) named 4-cyanopentanoic acid diselenobenzoate (RAFT-Se).
- As an oxidant in chemiluminescence reaction of anti-hypertensive drugs.
- As a component of the electrolyte solution for the cyclic voltammetry studies of phospholipid bilayers.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2
Supplementary Hazards
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service