Skip to Content
Merck
All Photos(1)

Key Documents

V800249

Sigma-Aldrich

Manganese(II) sulfate monohydrate

LR, 22-32% Mn basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnSO4 · H2O
CAS Number:
Molecular Weight:
169.02
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
powder
Pricing and availability is not currently available.

grade

LR

product line

Vetec

form

powder

concentration

22-32% Mn

pH

3.0-3.5 (20 °C, 50 g/L)

SMILES string

O.[Mn++].[O-]S([O-])(=O)=O

InChI

1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

ISPYRSDWRDQNSW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomás R Guilarte et al.
Neurotoxicology, 28(6), 1147-1152 (2007-07-31)
Humans exposed to excess levels of manganese (Mn(2+)) express psychiatric problems and deficits in attention and learning and memory. However, there is a paucity of knowledge on molecular mechanisms by which Mn(2+) produces such effects. We now report that Mn(2+)
Xiaojun Yang et al.
Animal science journal = Nihon chikusan Gakkaiho, 83(6), 474-481 (2012-06-15)
Iron is often found to be of excessive concentrations in laying hens' diets, which may cause antagonistic interactions with other minerals. This study was conducted to investigate how to supplement Zn and Mn in the diets without Fe supplementation. In
Edson R Silva et al.
Experimental parasitology, 125(2), 152-155 (2010-01-30)
In Leishmania, arginase is responsible for the production of ornithine, a precursor of polyamines required for proliferation of the parasite. In this work, the activation kinetics of immobilized arginase enzyme from L. (L.) amazonensis were studied by varying the concentration
David C Dorman et al.
Toxicological sciences : an official journal of the Society of Toxicology, 106(1), 46-54 (2008-08-08)
Neurotoxicity is linked with high-dose manganese inhalation. There are few biomarkers that correlate with manganese exposure. Blood manganese concentrations depend upon the magnitude and duration of the manganese exposure and inconsistently reflect manganese exposure concentrations. The objective of this study
Haixing Li et al.
Amino acids, 38(5), 1439-1445 (2009-09-30)
Production of gamma-aminobutyric acid (GABA) was carried out in Erlenmeyer flasks by Lactobacillus brevis NCL912. Traditional methods were first adopted to select the key factors that impact the GABA production to preliminarily determine the suitable concentration ranges of the key

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service