Skip to Content
Merck
All Photos(1)

Key Documents

V800232

Sigma-Aldrich

L-Ascorbic acid

AR, ≥99.5%

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

AR

product line

Vetec

Assay

≥99.5%

form

solid

mp

190-194 °C (dec.)

solubility

water: soluble 176 g/L at 20 °C

functional group

ester
hydroxyl
ketone

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Ascorbic acid or vitamin C is a water-soluble vitamin found in various foodstuffs. Benefits of vitamin C may include protection against immune system deficiencies, cardiovascular disease, hyperpigmentation, skin protection, anti-aging, iron absorption, and red blood cell production. It is also useful in the biosynthesis of collagen, L-carnitine, and neurotransmitters.

Application

L-ascorbic acid (L-AA) can be used as a reducing as well as a capping agent in the synthesis of well-dispersed metal nanoparticles and also in the reduction of graphene oxide sheets.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterization of gold nanoparticles using ascorbic acid as reducing agent in reverse micelles
Sun K, et al.
Journal of materials science & technology, 44(3), 754-758 (2009)
Jiali Zhang et al.
Chemical communications (Cambridge, England), 46(7), 1112-1114 (2010-02-04)
We demonstrated that the individual graphene oxide sheets can be readily reduced under a mild condition using L-ascorbic acid (L-AA). This simple approach should find practical applications in large scale production of water soluble graphene.
Vitamin C: One compound, several uses. Advances for delivery, efficiency and stability
Carita AC, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 31(2), 102117-102117 (2019)
Vitamin c
Linster CL and Van SE
FEBS Journal, 274(1), 1-22 (2007)
A review of micronutrients in sepsis: the role of thiamine, l-carnitine, vitamin C, selenium and vitamin D
Belsky JB, et al.
Nutrition Research Reviews, 31(2), 281-290 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service