Skip to Content
Merck
All Photos(1)

Key Documents

V800165

Sigma-Aldrich

Dimethylglyoxime

AR

Synonym(s):

2,3-Butanedione dioxime, Diacetyldioxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(=NOH)C(=NOH)CH3
CAS Number:
Molecular Weight:
116.12
Beilstein:
506731
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

AR

product line

Vetec

mp

240-241 °C (lit.)

functional group

amine
oxime

SMILES string

CC(=N/O)\C(C)=N\O

InChI

1S/C4H8N2O2/c1-3(5-7)4(2)6-8/h7-8H,1-2H3/b5-3+,6-4+

InChI key

JGUQDUKBUKFFRO-GGWOSOGESA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Capeness et al.
New biotechnology, 32(6), 727-731 (2015-02-18)
Desulfovibrio alaskensis G20 is an anaerobic sulfate reducing bacteria. While Desulfovibrio species have previously been shown to reduce palladium and platinum to the zero-state, forming nanoparticles in the process; there have been no reports that D. alaskensis is able to
Carsten R Hamann et al.
Contact dermatitis, 68(1), 15-22 (2012-12-12)
Nickel is widely used in coins; nickel may cause contact allergy and allergic contact dermatitis in those who handle them. To investigate alloy use, coin composition and nickel and cobalt release for a worldwide selection of currently circulating coins. Eight
Peter Jensen et al.
Contact dermatitis, 65(6), 354-358 (2011-11-15)
Despite the political intention to limit nickel allergy and dermatitis in Europeans, nickel allergy remains frequent. There are several explanations for the persistence of nickel allergy and dermatitis, including the increasing use of mobile phones. Before regulation of nickel release
Jandee Kim et al.
Langmuir : the ACS journal of surfaces and colloids, 27(23), 14638-14646 (2011-10-18)
Dimethylglyoxime (DMG) adsorbed on Au(111) was investigated using electrochemical scanning tunneling microscopy (STM) and X-ray photoelectron spectroscopy (XPS). STM experiments revealed three different structures of adsorbed DMG at open circuit potential (~0.07 V versus Ag/AgCl): (2√3×2√3)R30°-α, (2√3×4√3)R30°-β, and (2√3×4√3)R30°-γ. The
Jacob Pontoppidan Thyssen et al.
The Science of the total environment, 407(20), 5315-5318 (2009-07-28)
Nickel allergic subjects are at risk factor of acquiring hand eczema. In 1990 and 1994, respectively, Denmark and member states in the EU regulated nickel release from selected consumer products. The intention was that the nickel epidemic could be controlled

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service