All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
LR
form:
powder
grade
LR
vapor pressure
0.01 hPa ( 20 °C)
product line
Vetec™
Assay
≥97%
form
powder
pH
4-5 (5% in water)
mp
176 °C (dec.) (lit.)
SMILES string
O.O.Cl[Ca]Cl
InChI
1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2
InChI key
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sweta Rathore et al.
Journal of microencapsulation, 31(5), 469-478 (2014-04-05)
This work evaluated the feasibility of microencapsulating Clostridium acetobutylicum ATCC 824 cells by emulsification for fermentation to produce biobutanol. The effects of selected emulsification process on viability of the vegetative cells and spores were investigated to enable the selection of
P Geetha et al.
Ecotoxicology and environmental safety, 122, 17-23 (2015-07-15)
Population explosion, depletion of water resources and prolonged droughts and floods due to climatic change lead to scarcity of pure and hygienic drinking water in most of the developing countries. Recently nanomaterials attained considerable attention as biosorbent for water purification
Jiyoung Jung et al.
Colloids and surfaces. B, Biointerfaces, 104, 116-121 (2013-01-10)
The physical and chemical properties of commercial low methoxyl citrus pectins, CP 28 and CP 55, and a pectinmethylesterase (PME) charge modified citrus pectin (MP 38) were compared, and the differences in ability to encapsulate indomethacin in hydrogel beads was
Avinash A Kadam et al.
Bioresource technology, 132, 276-284 (2013-02-16)
Pretreatments to sugarcane bagasse (SCB) such as CaCl2, alkali, ammonia, steam and milling showed 91%, 46%, 47%, 42% and 56% adsorption of Solvent Red 5B (SR5B); 92%, 57%, 58%, 56% and 68% adsorption of simulated dyes mixture (SDM), and 86%
Chao-Han Lai et al.
Annals of surgery, 258(6), 1103-1110 (2013-01-09)
To investigate whether recombinant thrombomodulin containing all the extracellular domains (rTMD123) has therapeutic potential against aneurysm development. The pathogenesis of abdominal aortic aneurysm (AAA) is characterized by chronic inflammation and proteolytic degradation of extracellular matrix. Thrombomodulin, a transmembrane glycoprotein, exerts
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service