Skip to Content
Merck
All Photos(1)

Key Documents

V800082

Sigma-Aldrich

Sodium tetraborate decahydrate

AR, ≥99.5%

Synonym(s):

Borax decahydrate, Sodium borate decahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2B4O7 · 10H2O
CAS Number:
Molecular Weight:
381.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
AR
form:
crystalline powder
Pricing and availability is not currently available.

grade

AR

product line

Vetec

Assay

≥99.5%

form

crystalline powder

pH

9.0-10.0 (5% in water)

density

1.73 g/mL at 25 °C (lit.)

anion traces

carbonate (CO32-): ≤0.05%

cation traces

As: ≤0.005%

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B1Ob2ob([O-])ob(O1)o2

InChI

1S/B4O7.2Na.10H2O/c5-1-7-3-9-2(6)10-4(8-1)11-3;;;;;;;;;;;;/h;;;10*1H2/q-2;2*+1;;;;;;;;;;

InChI key

CDMADVZSLOHIFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eunice F S Vieira et al.
Biomacromolecules, 9(4), 1195-1199 (2008-03-19)
Oxidized alginate (ADA) and oxidized alginate blended with chitosan (ADA-Chit) were prepared in the presence of borax and CaCl 2, and their interactions with an antifolate drug, pyrimethamine (PYR), have been investigated. Tablets with a mean diameter of 1.2 +/-
Jian-Feng He et al.
Journal of chromatography. A, 1218(24), 3816-3821 (2011-05-14)
The paper presents an on-line transient moving chemical reaction boundary (MCRB) method for simply but efficiently stacking analytes in capillary electrophoresis (CE). The CE technique was developed for a rapid determination of fumaric and maleic acid. Based on the theory
Hasan Turkez
Journal of applied toxicology : JAT, 28(5), 658-664 (2007-11-21)
Titanium dioxide (TiO(2)) is a potential carcinogenic/mutagenic agent although it is used in many areas including medical industries and cosmetics. Boron (as boric acid and borax) has also well-described biological effects and therapeutic benefits. In a previous study, sister-chromatid exchanges
C Cencetti et al.
Carbohydrate polymers, 90(3), 1362-1370 (2012-09-04)
Silver-loaded dressings are designed to provide the same antimicrobial activity of topical silver, with the advantages of a sustained silver release and a reduced number of dressing changes. Moreover, such type of dressing must provide a moist environment, avoiding fiber
Marco Frasconi et al.
Langmuir : the ACS journal of surfaces and colloids, 25(18), 11097-11104 (2009-08-22)
A highly stable biological film was prepared by casting an aqueous dispersion of protein and composite hydrogel obtained from the polysaccharide Scleroglucan (Sclg) and borax as a cross-linking agent. Heme proteins, such as hemoglobin (Hb), myoglobin (Mb), and horseradish peroxidase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service