Skip to Content
Merck
All Photos(1)

Key Documents

V800071

Sigma-Aldrich

Benzaldehyde

LR, ≥99%

Synonym(s):

Bitter almond

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CHO
CAS Number:
Molecular Weight:
106.12
Beilstein:
471223
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
LR
bp:
178-179 °C (lit.)
vapor pressure:
4 mmHg ( 45 °C)

grade

LR

Quality Level

vapor density

3.7 (vs air)

vapor pressure

4 mmHg ( 45 °C)

product line

Vetec

Assay

≥99%

form

liquid

autoignition temp.

374 °F

expl. lim.

1.4 %, 20 °F

dilution

(for analytical testing)

refractive index

n20/D 1.545 (lit.)

pH

5.9 (20 °C)

bp

178-179 °C (lit.)

mp

−26 °C (lit.)

density

1.044 g/cm3 at 20 °C (lit.)

functional group

aldehyde
phenyl

SMILES string

O=Cc1ccccc1

InChI

1S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6H

InChI key

HUMNYLRZRPPJDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-gen Peng et al.
Chemical communications (Cambridge, England), 49(26), 2709-2711 (2013-02-26)
A center radially fibrous silica encapsulated TS-1 zeolite (TS-1@KCC-1) has been synthesized in a microemulsion system for the first time. Supporting the Rh(OH)3 species, this novel core-shell structured material serves as a robust bifunctional catalyst for one-pot synthesis of benzamide
Guanying Li et al.
Biomaterials, 63, 128-136 (2015-06-24)
Sulfur dioxide (SO2) and its derivatives sulfite and bisulfite play important roles in biological systems. However, in vivo detection of sulfite/bisulfite remains challenging. In this study, we developed a dinuclear Ir(III) complex (Ir4) as a two-photon phosphorescent probe for sulfite and
Catharine H Rankin
The Journal of experimental biology, 218(Pt 1), 41-49 (2015-01-09)
Inheritance of acquired characteristics without changes in DNA sequence has been called transgenerational epigenetics. This review looks at studies that used the model system Caenorhabditis elegans to uncover mechanisms of transgenerational epigenetics in studies of RNA interference, studies of longevity
Longyun Lv et al.
ACS combinatorial science, 15(4), 183-192 (2013-02-22)
We previously reported the novel efficient proton/heat-promoted four-component reactions (4CRs) of but-2-ynedioates, two same/different primary amines, and aldehydes for the synthesis of tetra- and pentasubstituted polyfunctional dihydropyrroles. If aromatic and aliphatic amines were used as reagents, four different series of
C Anitha et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 96, 493-500 (2012-06-26)
A series of metal(II) complexes of VO(II), Co(II), Ni(II), Cu(II) and Zn(II) have been synthesized from the azo Schiff base ligand 4-((E)-4-((E)-(4-chlorophenyl)diazenyl)-2-hydroxybenzylideneamino)-1,5-dimethyl-2-phenyl-1H-pyrazol-3(2H)-one (CDHBAP) and characterized by elemental analysis, spectral (IR, UV-Vis, (1)H NMR, ESR and EI-mass), magnetic moment measurements, molar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service