Skip to Content
Merck
All Photos(1)

Documents

V800018

Sigma-Aldrich

Acetic acid

suitable for HPLC, ≥99.8%

Synonym(s):

Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor density

2.07 (vs air)

product line

Vetec

Assay

≥99.8%

form

liquid

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

technique(s)

HPLC: suitable

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetic acid is an organic acid mainly used as a preservative, acidulating agent and flavoring agent. It is one of the main volatile constituents of vinegar and pyroligneous acid. Acetic acid is used as a flavoring agent in ice-creams, nonalcoholic beverages and baked goods. It is also used as a chemical reagent for the synthesis of vinyl acetate monomer, cellulose acetate, acetic anhydride, acetate ester, and terephthalic acid.

Application

Acetic acid can be used as a mobile phase additive to improve the separation and resolution of the bioactive compounds on the C18 column in reverse phase HPLC. It is also used as an analytical reference standard in chromatographic applications.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of pyroligneous acid from lignocellulosic biomass and their effectiveness against biological attacks
Lee SH, et al.
Journal of Applied Sciences Research, 10(20), 2440-2446 (2010)
Effect of mobile phase additives on the resolution of four bioactive compounds by RP-HPLC
Li, Shengnan, et al.
International Journal of Molecular Sciences, 11(5), 2229-2240 (2010)
Ice-cream as a probiotic food carrier
Cruz AG, et al.
Food Research International, 42(9), 1233-1239 (2009)
Organic acids and meat preservation: a review
Theron MM and Lues JFR
Food Reviews International, 23(2), 141-158 (2007)
Review of acetic acid synthesis from various feedstocks through different catalytic processes
Budiman AW, et al.
Catalysis Surveys from Asia, 20(3), 173-193 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service