Skip to Content
Merck
All Photos(1)

Key Documents

V800014

Sigma-Aldrich

2-Naphthol

LR, ≥98%

Synonym(s):

β-Naphthol, 2-Hydroxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OH
CAS Number:
Molecular Weight:
144.17
Beilstein:
742134
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

LR

vapor density

4.97 (vs air)

vapor pressure

10 mmHg ( 145.5 °C)

product line

Vetec

Assay

≥98%

form

solid

bp

285-286 °C (lit.)

mp

120-122 °C (lit.)

SMILES string

Oc1ccc2ccccc2c1

InChI

1S/C10H8O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11H

InChI key

JWAZRIHNYRIHIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

307.4 °F - closed cup

Flash Point(C)

153 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shouning Chai et al.
Environmental science & technology, 46(18), 10182-10190 (2012-08-28)
With in situ molecular imprinting technique, a novel nanoelectrode (MI, n-P)-TiO(2) with n-P heterojunction and molecular recognition ability was fabricated by liquid phase deposition at low temperature. Using bisphenol A (BPA) as template, the spindle-like TiO(2) particles 40-80 nm in
Hyung-Suk Yoon et al.
International journal of hygiene and environmental health, 215(4), 458-464 (2012-03-23)
Using the urinary biomarkers 1-hydroxypyrene-glucuronide (1-OHPG), 2-naphthol, and malondialdehyde (MDA), we evaluated seasonal and regional variations in polycyclic aromatic hydrocarbon (PAH) exposure and oxidative stress among Korean adults and children. In total, 322 children (175 male and 147 female) and
Takuya Oguma et al.
Journal of the American Chemical Society, 134(49), 20017-20020 (2012-11-28)
Iron(salan) complex 1 was found to catalyze the oxidative dearomatization of 1-substituted 2-naphthols with the formation of an all-carbon quaternary stereocenter in air in the presence of nitroalkanes, to afford the corresponding cyclic enones with high enantioselectivity of 88-96% ee.
Hyun-Hee Lim et al.
Food chemistry, 138(2-3), 791-796 (2013-02-16)
This paper describes a gas chromatography-mass spectrometric method of l-hydroxypyrene (1-HOP) and 2-naphthol (2-NAP) in fish and shellfish. Alkali hydrolysis method in this study was chosen and optimized to the reaction condition for 90 min at 90°C in a 2.0M
Glenn Talaska et al.
Toxicology letters, 213(1), 45-48 (2011-05-26)
Urinary bladder cancer is a historical disease of rubber workers often been associated with exposure to aromatic amines such as 2-naphthylamine. While exposure to these compounds has decreased markedly over time, the bladder cancer risk has not decreased in direct

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service