Skip to Content
Merck
All Photos(1)

Key Documents

V001331

Supelco

Hydroxylamine hydrochloride

99.0%

Synonym(s):

Hydroxylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2OH · HCl
CAS Number:
Molecular Weight:
69.49
Beilstein:
3539763
EC Number:
MDL number:
PubChem Substance ID:

Assay

99.0%

mp

155-157 °C (dec.) (lit.)

density

1.67 g/mL at 25 °C (lit.)

SMILES string

Cl.NO

InChI

1S/ClH.H3NO/c;1-2/h1H;2H,1H2

InChI key

WTDHULULXKLSOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

MAO inhibitor; inhibits platelet aggregation.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

spleen

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanderley R Bastos et al.
Environmental research, 140, 191-197 (2015-04-13)
The Madeira River is the largest tributary of the Amazon River Basin and one of the most impacted by artisanal gold-mining activities, deforestation for agricultural projects, and recent hydroelectric reservoirs. Total Hg (and methylmercury-MeHg) concentrations was determined in 3182 fish
Anke Bill et al.
PloS one, 9(6), e97973-e97973 (2014-06-03)
The human prostacyclin receptor (hIP receptor) is a seven-transmembrane G protein-coupled receptor (GPCR) that plays a critical role in vascular smooth muscle relaxation and platelet aggregation. hIP receptor dysfunction has been implicated in numerous cardiovascular abnormalities, including myocardial infarction, hypertension
Ayman M Atta et al.
International journal of molecular sciences, 16(4), 6911-6931 (2015-03-31)
In the present study, a new magnetic powder based on magnetite can be used as a petroleum crude oil collector. Amidoximes based on rosin as a natural product can be prepared from a reaction between hydroxylamine and rosin/acrylonitrile adducts. The
Yuan Zhao et al.
Small (Weinheim an der Bergstrasse, Germany), 10(22), 4770-4777 (2014-08-20)
Chiral plasmonic assemblies with strong and tunable chiroptical activity are emerging materials yet challenging to fabricate. Moreover, shell-programmed chiroptical regulation is really rare. Here, the chiroptical activity of core-shell (CS) nanoparticles (NPs) heterodimers (HDs) with different types and thicknesses of
Marte S Dragset et al.
Antimicrobial agents and chemotherapy, 59(4), 2256-2264 (2015-02-04)
Efficient iron acquisition is crucial for the pathogenesis of Mycobacterium tuberculosis. Mycobacterial iron uptake and metabolism are therefore attractive targets for antitubercular drug development. Resistance mutations against a novel pyrazolopyrimidinone compound (PZP) that is active against M. tuberculosis have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service