1681633
USP
Triclabendazole Related Compound B
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
5-Chloro-6-(2,3-dichlorophenoxy)-1,3-dihydro-2H-benzimidazole-2-thione
About This Item
Recommended Products
API family
triclabendazole
form
neat
packaging
pkg of 15 mg
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
InChI
1S/C13H7Cl3N2OS/c14-6-2-1-3-10(12(6)16)19-11-5-9-8(4-7(11)15)17-13(20)18-9/h1-5H,(H2,17,18,20)
InChI key
FUAVONSJNAMFPO-UHFFFAOYSA-N
General description
Application
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service