Skip to Content
Merck

Skip To

1663700

USP

Thiostrepton

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
200 mg
Check Cart for Availability
$526.00

About This Item

Empirical Formula (Hill Notation):
C72H85N19O18S5
CAS Number:
Molecular Weight:
1664.89
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107

$526.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


API family

thiostrepton

form

powder

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)
pharmaceutical

storage temp.

−20°C

InChI key

NSFFHOGKXHRQEW-AIHSUZKVSA-N

General description

Thiostrepton is a thiopeptide antibiotic produced by Streptomyces species, exhibiting bacteriostatic activity by binding to the 50S ribosomal subunit. It is primarily used in veterinary medicine. The USP standard enables assay validation and structural confirmation using chromatographic techniques.

The USP biologics antibiotics category includes a wide range of antimicrobial agents that are essential in treating bacterial infections. These antibiotics are derived from various natural sources or synthesized to combat specific pathogens effectively. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of antibiotic therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Thiostrepton USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Nystatin, Neomycin Sulfate, Thiostrepton, and Triamcinolone Acetonide Cream
  • Nystatin, Neomycin Sulfate, Thiostrepton, and Triamcinolone Acetonide Ointment

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of the USP Biologics program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
107630811024081097975
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)
pharmaceutical

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

form

powder

form

lyophilized powder (
)

form

-

form

-

API family

thiostrepton

API family

bleomycin

API family

cephapirin

API family

cefotetan

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

USP Biologics

USP Biologics

-

-


Still not finding the right product?

Explore all of our products under Thiostrepton


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Haga su pedido de una amplia gama de materiales de referencia primarios muy caracterizados para utilizar con las monografías de la USP-NF pensadas para el análisis de sustancias farmacológicas y formas de administración, excipientes farmacéuticos, ingredientes y suplementos alimentarios.

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.





Global Trade Item Number

SKUGTIN
1663700-200MG04061833833490

Questions

Reviews

No rating value

Active Filters