Skip to Content
Merck
All Photos(1)

Key Documents

1660000

USP

Thioguanine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

6-Thioguanine, 2-Amino-6-mercaptopurine, 2-Amino-6-purinethiol

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$512.00

$512.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
200 MG
$512.00

About This Item

Empirical Formula (Hill Notation):
C5H5N5S
CAS Number:
Molecular Weight:
167.19
Beilstein:
157765
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$512.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

thioguanine, tioguanine

manufacturer/tradename

USP

mp

≥300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NC1=Nc2nc[nH]c2C(=S)N1

InChI

1S/C5H5N5S/c6-5-9-3-2(4(11)10-5)7-1-8-3/h1H,(H4,6,7,8,9,10,11)

InChI key

WYWHKKSPHMUBEB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thioguanine USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Thioguanine Tablets

Biochem/physiol Actions

Ribosylated and phosphorylated by the same pathway as natural purine bases; as the nucleotide, inhibits a variety of cellular processes involved in nucleic acid synthesis. Has a long history as an effective treatment of leukemia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Di Matteo et al.
Clinical & experimental metastasis, 32(3), 289-300 (2015-02-05)
Tumor vessels are an attractive target for cancer therapy, including metastasis treatment. Angiogenesis inhibitors targeting the VEGF signalling pathway have proven to be efficacious in preclinical cancer models and in clinical trials. However, angiogenesis inhibition concomitantly elicits tumor adaptation and
Chenglong Sun et al.
Journal of immunology (Baltimore, Md. : 1950), 194(4), 1819-1831 (2015-01-18)
Herpesviruses are DNA viruses harboring the capacity to establish lifelong latent-recurrent infections. There is limited knowledge about viruses targeting the innate DNA-sensing pathway, as well as how the innate system impacts on the latent reservoir of herpesvirus infections. In this
Lynne Lennard et al.
British journal of haematology, 169(2), 228-240 (2014-12-03)
The impact of thiopurine methyltransferase (TPMT) genotype on thiopurine dose intensity, myelosuppression and treatment outcome was investigated in the United Kingdom childhood acute lymphoblastic leukaemia (ALL) trial ALL97. TPMT heterozygotes had significantly more frequent cytopenias and therefore required dose adjustments
Liang Liu et al.
Molecular cancer research : MCR, 12(11), 1597-1609 (2014-08-12)
Transforming growth factor beta (TGFβ) proteins are multitasking cytokines, in which high levels at tumor sites generally correlate with poor prognosis in human patients with cancer. Previously, it was reported that TGFβ downregulates the expression of ataxia telangiectasia-mutated (ATM) and
Brett P Gross et al.
The AAPS journal, 16(6), 1194-1203 (2014-09-17)
Metastatic breast cancer is currently incurable, and available therapies are associated with severe toxicities. Induction of protective anti-tumor immunity is a promising therapeutic approach for disseminated breast cancer, as immune responses are (i) systemic; (ii) antigen-specific; and (iii) capable of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service