Skip to Content
Merck

Skip To

1652012

USP

Tetrahydrozoline Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,2,3,4-Tetrahydronaphthaline-1-carbonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 mg
Check Cart for Availability
$1,630.00

About This Item

Empirical Formula (Hill Notation):
C11H11N
Molecular Weight:
157.21
NACRES:
NA.24
UNSPSC Code:
41116107

$1,630.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


API family

tetrahydrozoline

packaging

pkg of 50 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Tetrahydrozoline Related Compound A USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Tetrahydrozoline Hydrochloride
  • Tetrahydrozoline Hydrochloride Ophthalmic Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
165203416520231652056
application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

packaging

pkg of 50 mg

packaging

pkg of 25 mg

packaging

pkg of 25 mg

packaging

pkg of 25 mg

API family

tetrahydrozoline

API family

tetrahydrozoline

API family

tetrahydrozoline

API family

tetrahydrozoline


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1652012-50MG04065265964951

Questions

Reviews

No rating value

Active Filters