1614002
USP
Sodium fluoride
United States Pharmacopeia (USP) Reference Standard
About This Item
Recommended Products
grade
pharmaceutical primary standard
vapor pressure
1.4 mmHg ( 0 °C)
API family
sodium fluoride
manufacturer/tradename
USP
mp
993 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
[F-].[Na+]
InChI
1S/FH.Na/h1H;/q;+1/p-1
InChI key
PUZPDOWCWNUUKD-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Arresting Dental Caries with Silver Diamine Fluoride and Sodium Fluoride: A clinical trial investigates the efficacy of sequential applications of silver diamine fluoride gel followed by sodium fluoride varnish in arresting severe early childhood caries. The study demonstrates sodium fluoride′s critical role in dental health applications, particularly in enhancing the caries-arresting properties of treatments (Padilla Caceres TC et al., 2024).
- Mapping Vascular Microcalcification Using 18F-NaF PET/CT: This research uses sodium fluoride PET/CT scans to develop a spatial atlas for identifying and quantifying vascular microcalcifications in patients with hyperphosphatemic familial tumoral calcinosis. The findings highlight sodium fluoride′s application in advanced diagnostic imaging, providing a crucial tool for medical research and patient care (Sheppard AJ et al., 2024).
Analysis Note
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2
Supplementary Hazards
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service