Skip to Content
Merck
All Photos(1)

Key Documents

1612415

USP

Sesame Oil Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,2-Dilinoleoyl-3-oleoyl-rac-glycerol, 1,2-Di(cis-9,12-octadecadienoyl)-3-(cis-9-octadecenoyl)-rac-glycerol, OLL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H100O6
CAS Number:
Molecular Weight:
881.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

O=C(OC(COC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)CCCCCCC/C=C\C/C=C\CCCCC

InChI

1S/C57H100O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h16,18-19,21,25-30,54H,4-15,17,20,22-24,31-53H2,1-3H3/b19-16-,21-18-,28-25-,29-26-,30-27-

InChI key

VVEBTVMJPTZDHO-WECKWCTPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application


  • Sesamin in lipid metabolism research: A study in Biochemical and Biophysical Research Communications explored sesamin′s ability to alleviate lipid accumulation in liver cells, employing Sesame Oil Related Compound A as a biochemical analysis tool for observing molecular interactions within cellular pathways (Dong et al., 2024).

  • Characterization of lignans in black sesame: Research published in the Journal of Agricultural and Food Chemistry identified novel α-glucosidase inhibitors from sesame, utilizing Sesame Oil Related Compound A as a reference standard for quantifying and verifying the biochemical components of sesame derivatives (Doungwichitrkul et al., 2024).

  • Nanovesicle delivery systems from sesame: A study in Food Chemistry highlighted the use of exosome-like nanovesicles derived from sesame for luteolin delivery, showcasing the application of Sesame Oil Related Compound A in the stability and bioactivity testing of these organic chemistry components (Jiang et al., 2024).

  • Neuroprotective effects of sesamol: The Inflammopharmacology journal featured a study on sesamol′s protective effects against neuronal damage, where Sesame Oil Related Compound A was crucial for studying the compound′s interaction with cellular signaling pathways (El-Sayyad et al., 2024).

  • Sesamol and metabolic health: An investigation into the browning of white adipocytes through sesamol, detailed in The Journal of Nutritional Biochemistry, demonstrated the utility of Sesame Oil Related Compound A in examining hepatic signaling mechanisms and their influence on fat cells (Cheng et al., 2023).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service