Skip to Content
Merck

Skip To

1601441

USP

Residual Solvent Class 2 - Methylene chloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Dichloromethane solution, Methylene chloride in DMSO, Methylene chloride solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
3 x 1.2 mL
Check Cart for Availability
$576.00

About This Item

Linear Formula:
CH2Cl2
CAS Number:
Molecular Weight:
84.93
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

$576.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Residual Solvent Class 2 - Methylene chloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR15571.06049DX0838
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

liquid formulation
pharmaceutical

application(s)

-

format

neat

format

neat

format

-

format

-


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

188.6 °F

flash_point_c

87.00 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1601441-3X1.2ML04061838745125

Questions

Reviews

No rating value

Active Filters