Skip to Content
Merck
All Photos(1)

Key Documents

1568040

USP

Proguanil hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Chlorguanide, N1-(4-Chlorophenyl)-N5-isopropylbiguanide

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$447.00

$447.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

Select a Size

Change View
200 MG
$447.00

About This Item

Empirical Formula (Hill Notation):
C11H16ClN5·HCl
CAS Number:
Molecular Weight:
290.19
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$447.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

proguanil

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CC(C)NC(=N)NC(=N)Nc1ccc(Cl)cc1

InChI

1S/C11H16ClN5.ClH/c1-7(2)15-10(13)17-11(14)16-9-5-3-8(12)4-6-9;/h3-7H,1-2H3,(H5,13,14,15,16,17);1H

InChI key

SARMGXPVOFNNNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Proguanil hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Chlorguanide (proguanil) is combined with atovaquone for malaria prophylaxis. The two compounds act synergistically to inhibit the plasmodial dihydrofolate reductase (DHFR) and interrupt the electron transport chain. Mutations in DHFR account for the development of resistant strains.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rasmus Steen Pedersen et al.
Clinical and experimental pharmacology & physiology, 41(11), 870-878 (2014-08-15)
The aim of the present study was to determine the impact of CYP2C19*17 on the pharmacokinetics and pharmacodynamics of the active metabolite of clopidogrel and the pharmacokinetics of proguanil. Thus, we conducted an open-label two-phase cross-over study in 31 healthy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service