Skip to Content
Merck
All Photos(1)

Documents

1548407

USP

Potassium sorbate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Potassium 2,4-hexadienoate, Sorbic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCOOK
CAS Number:
Molecular Weight:
150.22
Beilstein:
5357554
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

potassium sorbate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[K+].C\C=C\C=C\C([O-])=O

InChI

1S/C6H8O2.K/c1-2-3-4-5-6(7)8;/h2-5H,1H3,(H,7,8);/q;+1/p-1/b3-2+,5-4+;

InChI key

CHHHXKFHOYLYRE-STWYSWDKSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Potassium sorbate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Budesonide Nasal Spray

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Budesonide Nasal Spray
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(3) (2020)
Potassium sorbate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(2), 5980-5980 (2020)
Jonathan Byrne et al.
Journal of pharmaceutical and biomedical analysis, 96, 111-117 (2014-04-16)
A novel stability-indicating reversed phase high performance liquid chromatographic (RP-HPLC) method for the simultaneous assay of betamethasone-17-valerate, fusidic acid and potassium sorbate as well as methyl- and propylparaben in a topical cream preparation has been developed. A 100mm×3.0mm ID. Ascentis
Ana Oliveira et al.
Food chemistry, 170, 74-83 (2014-10-13)
Strawberries purées are incorporated in foods and subjected to pH modulation according to the expected final food matrix. The effect of pH on strawberry polyphenols stored at 4 and 23 °C for 90 days was evaluated. Total antioxidant activity and
Andreas Lauterbach et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 88(3), 614-624 (2014-12-03)
A novel adapalene-loaded solid lipid microparticle (SLMA) dispersion as a topical drug delivery system (TDDS) for follicular penetration has been introduced. The objective of the present study was to investigate the rheological properties, the follicular penetration with differential tape stripping

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service