Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
50 mg | Check Cart for Availability | $660.00 |
About This Item
Empirical Formula (Hill Notation):
C20H11N2Na3O10S3
Molecular Weight:
604.47
NACRES:
NA.24
UNSPSC Code:
41116107
grade
pharmaceutical primary standard
agency
USP
API family
ponceau 4r
packaging
pkg of 50 mg
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
Ponceau 4R USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| application(s) pharmaceutical (small molecule) | application(s) - | application(s) cleaning products | application(s) diagnostic assay manufacturing |
| format neat | format - | format neat | format - |
| storage temp. 2-8°C | storage temp. -10 to -25°C | storage temp. - | storage temp. room temp |
| packaging pkg of 50 mg | packaging - | packaging - | packaging - |
| manufacturer/tradename USP | manufacturer/tradename Manufactured by: Sigma-Aldrich Production GmbH, Switzerland | manufacturer/tradename - | manufacturer/tradename - |
| API family ponceau 4r | API family - | API family - | API family - |
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

