Skip to Content
Merck
All Photos(1)

Key Documents

1535507

USP

Phenytoin sodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5,5-Diphenylhydantoin sodium salt, 5,5-Diphenyl-2,4-imidazolidinedione, Phenytoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H11N2O2Na
CAS Number:
Molecular Weight:
274.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

phenytoin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na]N1C(=O)NC(C1=O)(c2ccccc2)c3ccccc3

InChI

1S/C15H12N2O2.Na/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12;/h1-10H,(H2,16,17,18,19);/q;+1/p-1

InChI key

FJPYVLNWWICYDW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Phenytoin sodium is an anti-epileptic drug[1], which belongs to the hydantoin family.[2]

Application

Phenytoin sodium USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Reduces incidence of grand mal seizures; appears to stabilize excitable membranes perhaps through effects on Na+, K+, and Ca2+ channels.
Reduces incidence of grand mal seizures; appears to stabilize excitable membranes.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lemke.LT and Williams.AD
Foye's Principles of Medicinal Chemistry (2012)
Courtenay M and Griffiths M
Independent and Supplementary Prescribing: An Essential Guide null
Misty Good et al.
American journal of physiology. Gastrointestinal and liver physiology, 306(11), G1021-G1032 (2014-04-20)
Necrotizing enterocolitis (NEC) is the leading cause of death from gastrointestinal disease in premature infants and develops partly from an exaggerated intestinal epithelial immune response to indigenous microbes. There has been interest in administering probiotic bacteria to reduce NEC severity
Shareef Ahmed et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 30(10), 823-830 (2014-08-29)
To determine whether penetrating scleral or corneal injury can enhance intraocular penetration of systemic moxifloxacin, vancomycin, and ceftazidime. Thirty rabbits were divided into 3 groups for each antibiotic and then further subdivided to receive either scleral or corneal injury to
Maria B Rabaglino et al.
BMC genomics, 15, 1001-1001 (2014-11-21)
Major changes in gene expression occur in the fetal brain to modulate the function of this organ postnatally. Thus, factors can alter the genomics of the fetal brain, predisposing to neurological disorders later in life. We hypothesized that the physiological

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service