Skip to Content
Merck
All Photos(1)

Key Documents

1534209

USP

Phenyl salicylate melting point standard

approximately 41 degrees, United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Phenyl salicylate, Salol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$357.00

$357.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$357.00

About This Item

Linear Formula:
2-(HO)C6H4CO2C6H5
CAS Number:
Molecular Weight:
214.22
Beilstein:
393969
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$357.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

phenyl salicylate

quality

approximately 41 degrees

manufacturer/tradename

USP

bp

172-173 °C/12 mmHg (lit.)

mp

41-43 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1ccccc1C(=O)Oc2ccccc2

InChI

1S/C13H10O3/c14-12-9-5-4-8-11(12)13(15)16-10-6-2-1-3-7-10/h1-9,14H

InChI key

ZQBAKBUEJOMQEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenyl salicylate melting point standard may be used as a calorimetric reference material to investigate its melting behavior by studying its high-resolution specific enthalpy and specific heat capacity using adiabatic scanning calorimetry.[1]

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Investigation of the melting behavior of the reference materials biphenyl and phenyl salicylate by a new type adiabatic scanning calorimeter
Leys J, et al.
Thermochimica Acta, 582, 68-76 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service