Skip to Content
Merck
All Photos(1)

Documents

1499403

USP

Paricalcitol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(1α, 3β, 7E,22E)-19-Nor-9,10-secoergosta-5,7,22-triene-1,3,25-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

paricalcitol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

InChI

1S/C27H44O3/c1-18(8-9-19(2)26(3,4)30)24-12-13-25-21(7-6-14-27(24,25)5)11-10-20-15-22(28)17-23(29)16-20/h8-11,18-19,22-25,28-30H,6-7,12-17H2,1-5H3/b9-8+,21-11+/t18-,19+,22-,23-,24-,25+,27-/m1/s1

InChI key

BPKAHTKRCLCHEA-UBFJEZKGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Paricalcitol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Cozzolino et al.
Contributions to nephrology, 171, 161-165 (2011-06-01)
Cardiovascular (CV) morbidity and mortality are significantly higher in patients with chronic kidney disease (CKD). Mineral metabolism disorders, such as hyperphosphatemia, hypocalcemia, and vitamin D deficiency, have been deeply associated not only with bone disease, but also with vascular calcification
Julia A Lawrence et al.
Cancer biology & therapy, 14(6), 476-480 (2013-06-14)
The vitamin D hormone, [1,25(OH) 2D, calcitriol], inhibits proliferation and angiogenesis in breast cancer but its therapeutic use is limited by hypercalcemia. Synthetic analogs of 1,25(OH) 2D that are less calcemic, such as paricalcitol (19-nor-1,25-Dihydroxyvitamin D 2), are used to
Juan F Navarro-González et al.
Journal of clinical pharmacology, 53(4), 421-426 (2013-02-22)
Inflammation is a strong predictor of increased morbidity and mortality in hemodialysis (HD) patients. Paricalcitol, a selective vitamin D receptor activator used for prevention and treatment of secondary hyperparathyroidism, has shown anti-inflammatory properties in experimental studies, although clinical data are
Josefin Axelsson et al.
American journal of physiology. Renal physiology, 305(3), F237-F243 (2013-05-10)
Systemic infusions of ANG II rapidly induce large, dynamic increases in the permeability of the glomerular filtration barrier (GFB) in rats. After binding to its receptor(s), ANG II generates reactive oxygen species (ROS) and produces Ca²⁺ influx into cells, leading
Mario Cozzolino et al.
Current vascular pharmacology, 6(2), 148-153 (2008-04-09)
Hemodialysis (HD) patients are commonly affected by secondary hyperparathyroidism (SHPT), in which 3 well-known factors are usually involved: hypocalcemia, hyperphosphatemia and calcitriol deficiency. Classically, high parathyroid hormone (PTH) levels cause bone-associated diseases, such as osteitis fibrosa and renal osteodystrophy, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service