Skip to Content
Merck
All Photos(1)

Documents

1496959

USP

Paraldehyde

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,4,6-Trimethyl-1,3,5-trioxacyclohexane, s-Trimethyltrioxymethylene, Acetaldehyde trimer, NSC 9799, Paracetaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O3
CAS Number:
Molecular Weight:
132.16
Beilstein:
80142
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

paraldehyde

manufacturer/tradename

USP

density

0.994 g/mL at 20 °C (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

CC1OC(C)OC(C)O1

InChI

1S/C6H12O3/c1-4-7-5(2)9-6(3)8-4/h4-6H,1-3H3

InChI key

SQYNKIJPMDEDEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Reactant involved in:
  • Modified Sanford reaction
  • Synthesis of N-arylbenzoquinaldinium derivatives with antimicrobial activity
  • Hydroformylation of alkenylamines
  • Studies of molecules with inhibitory activity against HIV-1 integrase
  • Irregular Wittig reactions for the formation of α-hydroxyketones

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F

Flash Point(C)

24 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service