Skip to Content
Merck

Skip To

1470004

USP

Norethindrone acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

19-Norethindrone acetate, 17α-Ethynyl-17β-hydroxy-19-nor-4-androsten-3-one 17-acetate, 17α-Ethynyl-19-nortestosterone 17β-acetate, 17-Hydroxy-19-nor-17α-4-pregnen-20-yn-3-one acetate, 19-Norethisterone 17-acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
$576.00

About This Item

Empirical Formula (Hill Notation):
C22H28O3
CAS Number:
Molecular Weight:
340.46
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

$576.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

norethindrone

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C

InChI

1S/C22H28O3/c1-4-22(25-14(2)23)12-10-20-19-7-5-15-13-16(24)6-8-17(15)18(19)9-11-21(20,22)3/h1,13,17-20H,5-12H2,2-3H3/t17-,18+,19+,20-,21-,22-/m0/s1

InChI key

IMONTRJLAWHYGT-ZCPXKWAGSA-N

Gene Information

human ... PGR(5241)

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Norethindrone acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Estradiol and Norethindrone Acetate Tablets
  • Norethindrone Acetate
  • Norethindrone Acetate and Ethinyl Estradiol Tablets
  • Norethindrone Acetate Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0001532PHR2534N1225000
description

United States Pharmacopeia (USP) Reference Standard

description

European Pharmacopoeia (EP) Reference Standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

European Pharmacopoeia (EP) Reference Standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

EDQM

format

neat

format

neat

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

Gene Information

human ... PGR(5241)

Gene Information

human ... PGR(5241)

Gene Information

-

Gene Information

human ... PGR(5241)

API family

norethindrone

API family

norethisterone

API family

norethindrone

API family

norethisterone


Still not finding the right product?

Explore all of our products under Norethindrone acetate


Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters