1445550
USP
Monoclonal IgG System Suitability
United States Pharmacopeia (USP) Reference Standard
About This Item
Recommended Products
grade
pharmaceutical primary standard
form
lyophilized (cake)
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
−20°C
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
It is also used to prepare system suitability solution for the mobile phase as per the chapter 129 “Analytical procedures for recombinant therapeutic monoclonal antibodies” of United States Pharmacopeia (USP) for the measurement of:
- Low-molecular-weight species (LMWS, fragments) of monoclonal antibodies (mAbs) by size exclusion-high performance liquid chromatography (SE-HPLC)
- Product-related impurities such as non-glycosylated molecules, half antibodies, and fragments by capillary sodium dodecyl sulfate (CE-SDS) electrophoresis
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Discover our wide variety of products for intact mass analysis of monoclonal antibodies, including size-exclusion columns (SEC), ion exchange columns, reverse-phase columns, HPLC buffers, MALDI matrices and standards, high-purity solvents, reagents, tools for protein sample preparation, and certified reference materials.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service