Skip to Content
Merck

Skip To

1406005

USP

Methazolamide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5-Acetylimino-4-methyl-·2-1,3,4-thiadiazoline-2-sulfonamide, N-(4-Methyl-2-sulfamoyl-Δ2-1,3,4-thiadiazolin-5-ylidene) acetamide, L 584601, N-[5-(Aminosulfonyl)-3-methyl-1,3,4-thiadiazol-2(3H)-ylidene]acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Check Cart for Availability
$576.00

About This Item

Empirical Formula (Hill Notation):
C5H8N4O3S2
CAS Number:
Molecular Weight:
236.27
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

$576.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

methazolamide

manufacturer/tradename

USP

mp

208 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1N=C(S\C1=N\C(C)=O)S(N)(=O)=O

InChI

1S/C5H8N4O3S2/c1-3(10)7-4-9(2)8-5(13-4)14(6,11)12/h1-2H3,(H2,6,11,12)/b7-4+

InChI key

FLOSMHQXBMRNHR-QPJJXVBHSA-N

Gene Information

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methazolamide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Methazolamide Tablets

Biochem/physiol Actions

Cell-permeable and potent carbonic anhydrase (CA) inhibitor
Methazolamide is a cell-permeable and potent carbonic anhydrase (CA) inhibitor that is used in the treatment of glaucoma. Methazolamide is an insulin sensitizer that reduces hepatic glucose generation in animal models.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0720PHR32021005004
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

-

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

format

neat

format

-

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

Gene Information

human ... CA1(759), CA2(760), CA4(762), CA7(766)

Gene Information

human ... CA1(759), CA2(760), CA4(762), CA7(766)

Gene Information

-

Gene Information

human ... CA1(759), CA12(771), CA2(760), CA4(762)

mp

208 °C (dec.) (lit.)

mp

208 °C (dec.) (lit.)

mp

208 °C (dec.) (lit.)

mp

-


Still not finding the right product?

Explore all of our products under Methazolamide


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters