Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 mg | Check Cart for Availability | $1,690.00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
lindane
packaging
pkg of 25 mg
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
ClC1C(C(C(C(C1Cl)Cl)Cl)Cl)Cl
InChI
1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H
InChI key
JLYXXMFPNIAWKQ-UHFFFAOYSA-N
General description
Application
- Lindane
- Lindane Shampoo
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecules) |
| format neat | format neat | format neat | format neat |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| packaging pkg of 25 mg | packaging pkg of 20 mg | packaging pkg of 25 mg | packaging pkg of 25 mg |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| API family lindane | API family molindone | API family decitabine | API family nilotinib |
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




