Skip to Content
Merck
All Photos(1)

Key Documents

1366002

USP

Lidocaine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Diethylamino-N-(2,6-dimethylphenyl)acetamide, Lignocaine, Xylocaine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$569.00

$569.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$569.00

About This Item

Empirical Formula (Hill Notation):
C14H22N2O
CAS Number:
Molecular Weight:
234.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$569.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

lidocaine

manufacturer/tradename

USP

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCN(CC)CC(=O)Nc1c(C)cccc1C

InChI

1S/C14H22N2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4/h7-9H,5-6,10H2,1-4H3,(H,15,17)

InChI key

NNJVILVZKWQKPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lidocaine USP reference standard for use in specified quality tests and assays.

Also used to prepare standard and system suitability solution for assay, identification and impurity analysis according to given below monographs of the United States Pharmacopeia (USP):
  • Lidocaine[1]
  • Lidocaine Hydrochloride Topical Solution[2]
  • Lidocaine Hydrochloride Oral Topical Solution[3]
  • Lidocaine Hydrochloride Jelly[4]

Biochem/physiol Actions

Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration.
Na+ channel blocker; class IB antiarrhythmic.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lidocaine Hydrochloride Jelly
United States Pharmacopeia, 43(6), 2625-2625 (2020)
Lidocaine
United States Pharmacopeia, 43(3), 2620-2620 (2020)
Corrine A Nief et al.
Cancers, 14(19) (2022-10-15)
Ethanol ablation is a minimally invasive, cost-effective method of destroying tumor tissue through an intratumoral injection of high concentrations of cytotoxic alcohol. Ethyl-cellulose ethanol (ECE) ablation, a modified version of ethanol ablation, contains the phase-changing polysaccharide ethyl-cellulose to reduce ethanol
Lidocaine Hydrochloride Topical Solution
United States Pharmacopeia, 44(4), 2627-2627 (2020)
Lidocaine Hydrochloride Oral Topical Solution
United States Pharmacopeia, 44(4), 2626-2626 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service