Skip to Content
Merck
All Photos(1)

Key Documents

1345159

USP

Ioxaglic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-(2-Hydroxyethyl)-2,4,6-triiodo-5-{2-[2,4,6-triiodo-3-(N-methylacetamido)-5-(methylcarbamoyl)benzamido]acetamido}isophthalamic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$310.00

$310.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$310.00

About This Item

Empirical Formula (Hill Notation):
C24H21I6N5O8
CAS Number:
Molecular Weight:
1268.88
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$310.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

ioxaglic acid

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Ic1c(c(c(c(c1C(=O)NCC(=O)Nc2c(c(c(c(c2I)C(=O)O)I)C(=O)NCCO)I)I)C(=O)NC)I)N(C)C(=O)C

InChI

1S/C24H21I6N5O8/c1-7(37)35(3)20-17(29)10(21(39)31-2)13(25)11(18(20)30)23(41)33-6-8(38)34-19-15(27)9(22(40)32-4-5-36)14(26)12(16(19)28)24(42)43/h36H,4-6H2,1-3H3,(H,31,39)(H,32,40)(H,33,41)(H,34,38)(H,42,43)

InChI key

TYYBFXNZMFNZJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ioxaglic acid USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Ioxaglate Meglumine and Ioxaglate Sodium Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Delcour et al.
Cardiovascular and interventional radiology, 36(1), 237-238 (2012-06-15)
To test the compatibility of trisodium citrate, a catheter lock solution, with iodinated contrast medium. Iohexol, iobitridol, iodixanol, ioxaglate, ioxithalamate, iomeprol, and iopromide were tested. In all tests, 2 ml of contrast medium were mixed with 2 ml of trisodium citrate solution.
Laura Calvi et al.
Thyroid : official journal of the American Thyroid Association, 21(4), 443-449 (2011-03-10)
Thyrotoxicosis caused by destructive thyroiditis is self-limited and results from the subacute release of preformed thyroid hormone. Common etiologies include painful subacute thyroiditis and silent (painless) subacute thyroiditis (including postpartum thyroiditis, amiodarone-associated destructive thyroiditis, and lithium-associated thyroiditis). Thyrotoxicosis commonly evolves
G Kerckhofs et al.
European cells & materials, 25, 179-189 (2013-02-08)
We describe a non-destructive imaging method, named contrast-enhanced nanofocus X-ray computed tomography (CE-nanoCT), that permits simultaneously imaging and quantifying in 3D the (sub)tissue architecture and (biochemical) composition of cartilage and bone in small animal models at a novel contrast and
Emre Aslanger et al.
Coronary artery disease, 23(4), 265-270 (2012-02-22)
Contrast medium-induced nephropathy (CIN) is a well-known complication of coronary angiographic procedures, especially in patients treated with primary angioplasty. To prevent CIN, we examined using a local application of N-acetylcysteine (NAC) for the prevention of CIN during primary angioplasty. We
M Siebelt et al.
Osteoarthritis and cartilage, 19(10), 1183-1189 (2011-08-09)
Similar to delayed gadolinium enhanced MRI of cartilage, it might be possible to image cartilage quality using CT arthrography (CTa). This study assessed the potential of CTa as a clinically applicable tool to evaluate cartilage quality in terms of sulphated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service